Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615255 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf112 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf112 - chromosome 1 open reading frame 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320047.1 | Intron | NP_001306976.1 | ||||
NM_001320048.1 | Intron | NP_001306977.1 | ||||
NM_001320050.1 | Intron | NP_001306979.1 | ||||
NM_001320051.1 | Intron | NP_001306980.1 | ||||
NM_018186.3 | Intron | NP_060656.2 | ||||
XM_005245317.4 | Intron | XP_005245374.1 | ||||
XM_011509735.2 | Intron | XP_011508037.1 | ||||
XM_017001722.1 | Intron | XP_016857211.1 | ||||
XM_017001723.1 | Intron | XP_016857212.1 | ||||
XM_017001724.1 | Intron | XP_016857213.1 | ||||
XM_017001725.1 | Intron | XP_016857214.1 |
METTL18 - methyltransferase like 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |