Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603871 MIM: 123834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BYSL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BYSL - bystin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCND3 - cyclin D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136017.3 | Intron | NP_001129489.1 | ||||
NM_001136125.2 | Intron | NP_001129597.1 | ||||
NM_001136126.2 | Intron | NP_001129598.1 | ||||
NM_001287427.1 | Intron | NP_001274356.1 | ||||
NM_001287434.1 | Intron | NP_001274363.1 | ||||
NM_001760.4 | Intron | NP_001751.1 | ||||
XM_011514971.1 | Intron | XP_011513273.1 | ||||
XM_011514972.1 | Intron | XP_011513274.1 |