Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NAA11 - N(alpha)-acetyltransferase 11, NatA catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032693.2 | 197 | Missense Mutation | ACT,GCT | T,A 6 | NP_116082.1 | |
XM_017008777.1 | 197 | Missense Mutation | ACT,GCT | T,A 6 | XP_016864266.1 | |
XM_017008778.1 | 197 | Missense Mutation | ACT,GCT | T,A 6 | XP_016864267.1 | |
XM_017008779.1 | 197 | Missense Mutation | ACT,GCT | T,A 6 | XP_016864268.1 |
Set Membership: |
HapMap |