Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCTGGGCCACCTCTGCCCCCC[A/C]CATTCTGGTCAGAATGAGCCTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104620 MIM: 601832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD14A-ACY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD14A-ACY1 - ABHD14A-ACY1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ACY1 - aminoacylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL29 - ribosomal protein L29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000992.2 | Intron | NP_000983.1 |