Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605949 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRC5C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPRC5C - G protein-coupled receptor class C group 5 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018653.3 | 108 | Intron | NP_061123.3 | |||
NM_022036.2 | 108 | Intron | NP_071319.2 | |||
XM_005257512.1 | 108 | Intron | XP_005257569.1 | |||
XM_005257514.1 | 108 | UTR 5 | XP_005257571.1 | |||
XM_017024855.1 | 108 | Intron | XP_016880344.1 | |||
XM_017024856.1 | 108 | Intron | XP_016880345.1 |
Set Membership: |
HapMap |