Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613539 MIM: 604789 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FEM1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FEM1B - fem-1 homolog B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGA11 - integrin subunit alpha 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004439.1 | 3792 | UTR 3 | NP_001004439.1 | |||
XM_005254228.3 | 3792 | UTR 3 | XP_005254285.1 | |||
XM_011521363.2 | 3792 | UTR 3 | XP_011519665.1 |
Set Membership: |
HapMap |