Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300482 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GAB3 - GRB2 associated binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081573.2 | 3994 | UTR 3 | NP_001075042.1 | |||
NM_001282283.1 | 3994 | UTR 3 | NP_001269212.1 | |||
NM_080612.3 | 3994 | UTR 3 | NP_542179.1 | |||
XM_005274648.1 | 3994 | Intron | XP_005274705.1 | |||
XM_006724804.1 | 3994 | UTR 3 | XP_006724867.1 | |||
XM_011531103.1 | 3994 | UTR 3 | XP_011529405.1 | |||
XM_011531105.1 | 3994 | Intron | XP_011529407.1 | |||
XM_011531106.1 | 3994 | UTR 3 | XP_011529408.1 | |||
XM_017029276.1 | 3994 | UTR 3 | XP_016884765.1 |
Set Membership: |
HapMap |