Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107272 MIM: 601782 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD72 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD72 - CD72 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001782.2 | 1690 | UTR 3 | NP_001773.1 | |||
XM_006716893.2 | 1690 | UTR 3 | XP_006716956.1 | |||
XM_006716894.3 | 1690 | UTR 3 | XP_006716957.1 | |||
XM_011518074.2 | 1690 | UTR 3 | XP_011516376.1 | |||
XM_017015351.1 | 1690 | UTR 3 | XP_016870840.1 |
MIR4667 - microRNA 4667 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TESK1 - testis-specific kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318230.1 | 1690 | Intron | NP_001305159.1 | |||
NM_006285.2 | 1690 | Intron | NP_006276.2 |