Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGTCCAATAGCTCAAGTTTAC[A/G]AAGTACGGAATGTAAATATCTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603565 MIM: 614784 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNT4 - polypeptide N-acetylgalactosaminyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003774.4 | 2655 | UTR 3 | NP_003765.2 |
POC1B - POC1 centriolar protein B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199777.1 | 2655 | Intron | NP_001186706.1 | |||
NM_172240.2 | 2655 | Intron | NP_758440.1 |
POC1B-GALNT4 - POC1B-GALNT4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199781.1 | 2655 | UTR 3 | NP_001186710.1 | |||
NM_001199782.1 | 2655 | UTR 3 | NP_001186711.1 |
Set Membership: |
HapMap |