Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609073 MIM: 611585 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXW8 - F-box and WD repeat domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TESC - tescalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168325.1 | Intron | NP_001161797.1 | ||||
NM_017899.3 | Intron | NP_060369.3 | ||||
XM_011538503.1 | Intron | XP_011536805.1 | ||||
XM_017019532.1 | Intron | XP_016875021.1 | ||||
XM_017019533.1 | Intron | XP_016875022.1 |