Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCAGAAGACCCCCAGGCGGCCA[C/T]CCCGCATGGTTGTGTCCAAGACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600310 MIM: 607536 | ||||||||||||||||||||
Literature Links: |
COMP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COMP - cartilage oligomeric matrix protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000095.2 | 2192 | Missense Mutation | GAT,GGT | D,G 719 | NP_000086.2 |
CRTC1 - CREB regulated transcription coactivator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098482.1 | 2192 | Intron | NP_001091952.1 | |||
NM_015321.2 | 2192 | Intron | NP_056136.2 | |||
XM_005259833.3 | 2192 | Intron | XP_005259890.1 | |||
XM_005259834.1 | 2192 | Intron | XP_005259891.1 | |||
XM_005259835.3 | 2192 | Intron | XP_005259892.1 | |||
XM_005259836.3 | 2192 | Intron | XP_005259893.1 | |||
XM_006722710.3 | 2192 | Intron | XP_006722773.1 | |||
XM_011527842.2 | 2192 | Intron | XP_011526144.1 | |||
XM_017026536.1 | 2192 | Intron | XP_016882025.1 | |||
XM_017026537.1 | 2192 | Intron | XP_016882026.1 | |||
XM_017026538.1 | 2192 | Intron | XP_016882027.1 |