Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCAGGCAGGTCACGGGGGCATG[C/T]TGTGCTGCCAAGTCCTCCAAAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613842 MIM: 611270 | ||||||||||||||||||||
Literature Links: |
GZF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GZF1 - GDNF inducible zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317012.1 | 3735 | Intron | NP_001303941.1 | |||
NM_001317019.1 | 3735 | Intron | NP_001303948.1 | |||
NM_022482.4 | 3735 | Intron | NP_071927.1 |
NAPB - NSF attachment protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283018.1 | 3735 | UTR 3 | NP_001269947.1 | |||
NM_001283020.1 | 3735 | UTR 3 | NP_001269949.1 | |||
NM_001283026.1 | 3735 | UTR 3 | NP_001269955.1 | |||
NM_022080.2 | 3735 | UTR 3 | NP_071363.1 |