Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACACAGACTTTTTAAAGAAAGC[A/G]GAGTGAAAATCAACTATGTGTACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605787 MIM: 616194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRA2 - ankyrin repeat family A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP15 - UTP15, small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284430.1 | Intron | NP_001271359.1 | ||||
NM_001284431.1 | Intron | NP_001271360.1 | ||||
NM_032175.3 | Intron | NP_115551.2 | ||||
XM_011543680.2 | Intron | XP_011541982.1 |