Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGGCGTCTTCGAATGCGGCCTA[A/G]GGCGCCTGCCGCCAGTCTCCTGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C17orf53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf53 - chromosome 17 open reading frame 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171251.2 | 56 | UTR 5 | NP_001164722.1 | |||
NM_001321310.1 | 56 | UTR 5 | NP_001308239.1 | |||
NM_001321311.1 | 56 | UTR 5 | NP_001308240.1 | |||
NM_024032.4 | 56 | UTR 5 | NP_076937.2 |