Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGACCCCACCCCGTTCACAGG[T/C]CCGGCCCACGAGAGGCCAATGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTLL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTLL10 - tubulin tyrosine ligase like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130045.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | NP_001123517.1 | |
NM_153254.2 | 511 | UTR 5 | NP_694986.2 | |||
XM_005244738.1 | 511 | UTR 5 | XP_005244795.1 | |||
XM_011541177.2 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_011539479.1 | |
XM_017000906.1 | 511 | Silent Mutation | GGC,GGT | G,G 40 | XP_016856395.1 | |
XM_017000907.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_016856396.1 | |
XM_017000908.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_016856397.1 | |
XM_017000909.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_016856398.1 | |
XM_017000910.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_016856399.1 | |
XM_017000911.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_016856400.1 | |
XM_017000912.1 | 511 | Silent Mutation | GGC,GGT | G,G 67 | XP_016856401.1 |
TTLL10-AS1 - TTLL10 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |