Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGACTTTAAAAGTAAGTTGTTT[A/G]CTAATTTCTGGACTAATTATGCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608433 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC339529 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC339529 - uncharacterized LOC339529 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB18 - zinc finger and BTB domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278196.1 | 8 | Intron | NP_001265125.1 | |||
NM_006352.4 | 8 | UTR 5 | NP_006343.2 | |||
NM_205768.2 | 8 | Intron | NP_991331.1 | |||
XM_005273006.2 | 8 | Intron | XP_005273063.1 | |||
XM_017000060.1 | 8 | Intron | XP_016855549.1 |