Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTACTAGTAGACAAGATATC[C/T]ACTAAACTCCAAGTAATTAAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf50 - chromosome 12 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152589.1 | Intron | NP_689802.1 | ||||
XM_011537985.1 | Intron | XP_011536287.1 | ||||
XM_017018886.1 | Intron | XP_016874375.1 | ||||
XM_017018887.1 | Intron | XP_016874376.1 | ||||
XM_017018888.1 | Intron | XP_016874377.1 |