Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGCAGTGGAGTGATTGACGTGC[C/T]TGAGTTTGAAGAGAGTTAACCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDEL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDEL1 - nudE neurodevelopment protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025579.2 | 313 | Intron | NP_001020750.1 | |||
NM_030808.4 | 313 | Intron | NP_110435.1 | |||
XM_011524013.1 | 313 | Intron | XP_011522315.1 | |||
XM_017025183.1 | 313 | UTR 5 | XP_016880672.1 | |||
XM_017025184.1 | 313 | UTR 5 | XP_016880673.1 | |||
XM_017025185.1 | 313 | Intron | XP_016880674.1 | |||
XM_017025186.1 | 313 | UTR 5 | XP_016880675.1 | |||
XM_017025187.1 | 313 | Intron | XP_016880676.1 | |||
XM_017025188.1 | 313 | UTR 5 | XP_016880677.1 |