Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTGGCTTCCCTCCTCTCCCGAA[A/G]AAAGGGCTCTGGAGGGGCTCCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164831 MIM: 616700 | ||||||||||||||||||||
Literature Links: |
BMI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMI1 - BMI1 proto-oncogene, polycomb ring finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005180.8 | Intron | NP_005171.4 |
COMMD3 - COMM domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COMMD3-BMI1 - COMMD3-BMI1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204062.1 | Intron | NP_001190991.1 |