Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTGACTTTGAAAGGAAAAGGG[A/G]ACTGAGAGAGCCCCCAGACGCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC17A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC17A - C-type lectin domain family 17 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204118.1 | 181 | Intron | NP_001191047.1 | |||
NM_207390.3 | 181 | Intron | NP_997273.3 | |||
XM_017026785.1 | 181 | UTR 5 | XP_016882274.1 | |||
XM_017026786.1 | 181 | UTR 5 | XP_016882275.1 | |||
XM_017026787.1 | 181 | UTR 5 | XP_016882276.1 | |||
XM_017026788.1 | 181 | Intron | XP_016882277.1 | |||
XM_017026789.1 | 181 | Intron | XP_016882278.1 | |||
XM_017026790.1 | 181 | UTR 5 | XP_016882279.1 | |||
XM_017026791.1 | 181 | UTR 5 | XP_016882280.1 | |||
XM_017026792.1 | 181 | UTR 5 | XP_016882281.1 | |||
XM_017026793.1 | 181 | UTR 5 | XP_016882282.1 | |||
XM_017026794.1 | 181 | UTR 5 | XP_016882283.1 |