Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAAGGAATAGTGTTAGATTCTG[A/G]ATTAAGATGAAAGCGTATCGATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603015 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3609 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3609 - microRNA 3609 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA28 - small Cajal body-specific RNA 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRRAP - transformation/transcription domain associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244580.1 | Intron | NP_001231509.1 | ||||
NM_003496.3 | Intron | NP_003487.1 |