Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACTCCACGCCTTTCTTCATTTA[C/T]CCTTCAAAATGGCGGACTTTAGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM3A - lysine demethylase 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146688.1 | 34 | Intron | NP_001140160.1 | |||
NM_018433.5 | 34 | Intron | NP_060903.2 | |||
XM_006712051.3 | 34 | Intron | XP_006712114.1 | |||
XM_017004493.1 | 34 | UTR 5 | XP_016859982.1 | |||
XM_017004494.1 | 34 | Intron | XP_016859983.1 |