Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACCCAAAGGACTCTAATTATGG[A/G]AATTCTAGAGATCATATGAAACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603433 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF143 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF143 - zinc finger protein 143 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282656.1 | Intron | NP_001269585.1 | ||||
NM_001282657.1 | Intron | NP_001269586.1 | ||||
NM_003442.5 | Intron | NP_003433.3 | ||||
XM_005253121.1 | Intron | XP_005253178.1 | ||||
XM_005253122.1 | Intron | XP_005253179.1 | ||||
XM_005253127.1 | Intron | XP_005253184.1 | ||||
XM_011520349.2 | Intron | XP_011518651.1 | ||||
XM_017018252.1 | Intron | XP_016873741.1 | ||||
XM_017018253.1 | Intron | XP_016873742.1 | ||||
XM_017018254.1 | Intron | XP_016873743.1 | ||||
XM_017018255.1 | Intron | XP_016873744.1 | ||||
XM_017018256.1 | Intron | XP_016873745.1 |