Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTTGGAAAGGGTTTTGAAGGGGG[C/T]TCTAAGGGGGAGTGGGCGCCTAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100131655 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100131655 - uncharacterized LOC100131655 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF236 - zinc finger protein 236 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306089.1 | Intron | NP_001293018.1 | ||||
NM_007345.3 | Intron | NP_031371.3 | ||||
XM_005266762.3 | Intron | XP_005266819.1 | ||||
XM_011526165.2 | Intron | XP_011524467.1 | ||||
XM_011526166.2 | Intron | XP_011524468.1 | ||||
XM_011526168.2 | Intron | XP_011524470.1 | ||||
XM_011526169.2 | Intron | XP_011524471.1 |