Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCCAGGTACACATGACATTCAC[A/G]CCCTGATCCTTGGGAGAGCTATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608801 MIM: 611487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCDH - glutaryl-CoA dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000159.3 | 1369 | Missense Mutation | ACC,GCC | T,A 421 | NP_000150.1 | |
NM_013976.3 | 1369 | Intron | NP_039663.1 | |||
XM_006722721.2 | 1369 | Intron | XP_006722784.1 | |||
XM_011527899.2 | 1369 | Intron | XP_011526201.1 | |||
XM_011527900.2 | 1369 | Intron | XP_011526202.1 | |||
XM_017026580.1 | 1369 | Intron | XP_016882069.1 |
SYCE2 - synaptonemal complex central element protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105578.1 | 1369 | Intron | NP_001099048.1 | |||
XM_005259848.4 | 1369 | Intron | XP_005259905.1 | |||
XM_011527882.2 | 1369 | Intron | XP_011526184.1 | |||
XM_011527883.2 | 1369 | Intron | XP_011526185.1 |