Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600925 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRJ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPRJ - protein tyrosine phosphatase, receptor type J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098503.1 | Intron | NP_001091973.1 | ||||
NM_002843.3 | Intron | NP_002834.3 | ||||
XM_017018083.1 | Intron | XP_016873572.1 | ||||
XM_017018084.1 | Intron | XP_016873573.1 | ||||
XM_017018085.1 | Intron | XP_016873574.1 |
Set Membership: |
HapMap |