Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610179 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLB1 - phospholipase B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170585.1 | 88 | Intron | NP_001164056.1 | |||
NM_153021.4 | 88 | Intron | NP_694566.4 | |||
XM_011532579.2 | 88 | UTR 5 | XP_011530881.1 | |||
XM_011532581.2 | 88 | UTR 5 | XP_011530883.1 | |||
XM_011532584.2 | 88 | UTR 5 | XP_011530886.1 | |||
XM_011532588.2 | 88 | UTR 5 | XP_011530890.1 | |||
XM_011532589.2 | 88 | UTR 5 | XP_011530891.1 | |||
XM_011532590.2 | 88 | UTR 5 | XP_011530892.1 | |||
XM_011532591.2 | 88 | UTR 5 | XP_011530893.1 | |||
XM_011532593.2 | 88 | Intron | XP_011530895.1 | |||
XM_011532601.2 | 88 | Intron | XP_011530903.1 | |||
XM_011532602.2 | 88 | Intron | XP_011530904.1 | |||
XM_011532603.2 | 88 | Intron | XP_011530905.1 | |||
XM_011532606.2 | 88 | UTR 5 | XP_011530908.1 | |||
XM_011532607.2 | 88 | UTR 5 | XP_011530909.1 | |||
XM_011532608.2 | 88 | UTR 5 | XP_011530910.1 | |||
XM_011532609.2 | 88 | Intron | XP_011530911.1 | |||
XM_011532610.2 | 88 | Intron | XP_011530912.1 | |||
XM_017003432.1 | 88 | UTR 5 | XP_016858921.1 | |||
XM_017003433.1 | 88 | UTR 5 | XP_016858922.1 | |||
XM_017003434.1 | 88 | UTR 5 | XP_016858923.1 |
Set Membership: |
HapMap |