Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCCAAGCTCAGAATTCTCTCC[C/T]TCCTCTTTGGTCTAATCTCCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613469 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPSE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HPSE2 - heparanase 2 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166244.1 | 3228 | UTR 3 | NP_001159716.1 | |||
NM_001166245.1 | 3228 | UTR 3 | NP_001159717.1 | |||
NM_001166246.1 | 3228 | UTR 3 | NP_001159718.1 | |||
NM_021828.4 | 3228 | UTR 3 | NP_068600.4 | |||
XM_006717937.2 | 3228 | UTR 3 | XP_006718000.1 | |||
XM_011540029.1 | 3228 | UTR 3 | XP_011538331.1 | |||
XM_011540030.1 | 3228 | UTR 3 | XP_011538332.1 | |||
XM_011540031.2 | 3228 | UTR 3 | XP_011538333.1 | |||
XM_011540033.2 | 3228 | UTR 3 | XP_011538335.1 | |||
XM_017016495.1 | 3228 | Intron | XP_016871984.1 | |||
XM_017016496.1 | 3228 | Intron | XP_016871985.1 | |||
XM_017016497.1 | 3228 | UTR 3 | XP_016871986.1 | |||
XM_017016498.1 | 3228 | UTR 3 | XP_016871987.1 |
LOC101927278 - uncharacterized LOC101927278 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |