Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607189 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RGS8 - regulator of G-protein signaling 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102450.2 | Intron | NP_001095920.1 | ||||
NM_033345.3 | Intron | NP_203131.1 | ||||
XM_005245555.3 | Intron | XP_005245612.1 | ||||
XM_011510089.2 | Intron | XP_011508391.1 | ||||
XM_011510090.2 | Intron | XP_011508392.1 | ||||
XM_011510091.2 | Intron | XP_011508393.1 | ||||
XM_017002631.1 | Intron | XP_016858120.1 | ||||
XM_017002632.1 | Intron | XP_016858121.1 | ||||
XM_017002633.1 | Intron | XP_016858122.1 | ||||
XM_017002634.1 | Intron | XP_016858123.1 | ||||
XM_017002635.1 | Intron | XP_016858124.1 | ||||
XM_017002636.1 | Intron | XP_016858125.1 | ||||
XM_017002637.1 | Intron | XP_016858126.1 |
Set Membership: |
HapMap |