Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTBN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPTBN1 - spectrin beta, non-erythrocytic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003128.2 | Intron | NP_003119.2 | ||||
NM_178313.2 | Intron | NP_842565.2 | ||||
XM_005264517.2 | Intron | XP_005264574.1 | ||||
XM_005264518.1 | Intron | XP_005264575.1 | ||||
XM_006712087.2 | Intron | XP_006712150.1 | ||||
XM_017004779.1 | Intron | XP_016860268.1 | ||||
XM_017004780.1 | Intron | XP_016860269.1 | ||||
XM_017004781.1 | Intron | XP_016860270.1 |