Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614139 MIM: 608998 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAPPC12 - trafficking protein particle complex 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321102.1 | Intron | NP_001308031.1 | ||||
NM_016030.5 | Intron | NP_057114.5 | ||||
XM_011510350.2 | Intron | XP_011508652.1 | ||||
XM_011510352.2 | Intron | XP_011508654.1 | ||||
XM_011510353.2 | Intron | XP_011508655.1 | ||||
XM_011510354.2 | Intron | XP_011508656.1 | ||||
XM_011510355.2 | Intron | XP_011508657.1 | ||||
XM_011510356.1 | Intron | XP_011508658.1 | ||||
XM_017004257.1 | Intron | XP_016859746.1 |
TSSC1 - tumor suppressing subtransferable candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |