Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF619 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF619 - zinc finger protein 619 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145082.2 | 247 | UTR 5 | NP_001138554.1 | |||
NM_001145083.1 | 247 | UTR 5 | NP_001138555.1 | |||
NM_001145093.2 | 247 | UTR 5 | NP_001138565.1 | |||
XM_006713112.2 | 247 | UTR 5 | XP_006713175.1 | |||
XM_006713113.3 | 247 | UTR 5 | XP_006713176.1 | |||
XM_011533607.1 | 247 | UTR 5 | XP_011531909.1 | |||
XM_011533608.1 | 247 | UTR 5 | XP_011531910.1 | |||
XM_017006225.1 | 247 | Intron | XP_016861714.1 |