Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TKT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TKT - transketolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001064.3 | 1924 | Missense Mutation | AAG,AGG | K,R 597 | NP_001055.1 | |
NM_001135055.2 | 1924 | Missense Mutation | AAG,AGG | K,R 597 | NP_001128527.1 | |
NM_001258028.1 | 1924 | Missense Mutation | AAG,AGG | K,R 605 | NP_001244957.1 | |
XM_011534054.1 | 1924 | Missense Mutation | AAG,AGG | K,R 605 | XP_011532356.1 | |
XM_011534055.1 | 1924 | Missense Mutation | AAG,AGG | K,R 431 | XP_011532357.1 |