Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606347 | ||||||||||||||||||||
Literature Links: |
PSTPIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSTPIP1 - proline-serine-threonine phosphatase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321135.1 | Intron | NP_001308064.1 | ||||
NM_001321136.1 | Intron | NP_001308065.1 | ||||
NM_001321137.1 | Intron | NP_001308066.1 | ||||
NM_003978.4 | Intron | NP_003969.2 | ||||
XM_006720737.2 | Intron | XP_006720800.1 | ||||
XM_011522163.1 | Intron | XP_011520465.1 | ||||
XM_011522164.1 | Intron | XP_011520466.1 | ||||
XM_011522165.2 | Intron | XP_011520467.1 | ||||
XM_011522166.1 | Intron | XP_011520468.1 | ||||
XM_011522167.1 | Intron | XP_011520469.1 | ||||
XM_011522168.2 | Intron | XP_011520470.1 | ||||
XM_011522169.1 | Intron | XP_011520471.1 |