Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194554 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF45 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF45 - zinc finger protein 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003425.3 | 3542 | UTR 3 | NP_003416.1 | |||
XM_011527267.1 | 3542 | UTR 3 | XP_011525569.1 | |||
XM_011527269.1 | 3542 | UTR 3 | XP_011525571.1 | |||
XM_011527271.1 | 3542 | UTR 3 | XP_011525573.1 | |||
XM_011527273.1 | 3542 | UTR 3 | XP_011525575.1 | |||
XM_017027217.1 | 3542 | UTR 3 | XP_016882706.1 | |||
XM_017027218.1 | 3542 | UTR 3 | XP_016882707.1 | |||
XM_017027219.1 | 3542 | UTR 3 | XP_016882708.1 | |||
XM_017027220.1 | 3542 | UTR 3 | XP_016882709.1 | |||
XM_017027221.1 | 3542 | UTR 3 | XP_016882710.1 | |||
XM_017027222.1 | 3542 | UTR 3 | XP_016882711.1 | |||
XM_017027223.1 | 3542 | UTR 3 | XP_016882712.1 | |||
XM_017027224.1 | 3542 | UTR 3 | XP_016882713.1 | |||
XM_017027225.1 | 3542 | UTR 3 | XP_016882714.1 | |||
XM_017027226.1 | 3542 | UTR 3 | XP_016882715.1 | |||
XM_017027227.1 | 3542 | UTR 3 | XP_016882716.1 | |||
XM_017027228.1 | 3542 | UTR 3 | XP_016882717.1 | |||
XM_017027229.1 | 3542 | UTR 3 | XP_016882718.1 | |||
XM_017027230.1 | 3542 | UTR 3 | XP_016882719.1 |
Set Membership: |
HapMap |