Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611054 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR548K - microRNA 548k | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPFIA1 - PTPRF interacting protein alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003626.3 | Intron | NP_003617.1 | ||||
NM_177423.2 | Intron | NP_803172.1 | ||||
XM_006718716.3 | Intron | XP_006718779.1 | ||||
XM_011545306.2 | Intron | XP_011543608.1 | ||||
XM_011545307.2 | Intron | XP_011543609.1 | ||||
XM_011545308.2 | Intron | XP_011543610.1 | ||||
XM_011545309.2 | Intron | XP_011543611.1 | ||||
XM_011545310.2 | Intron | XP_011543612.1 | ||||
XM_011545311.2 | Intron | XP_011543613.1 | ||||
XM_011545312.2 | Intron | XP_011543614.1 | ||||
XM_011545313.2 | Intron | XP_011543615.1 | ||||
XM_011545314.2 | Intron | XP_011543616.1 | ||||
XM_011545315.2 | Intron | XP_011543617.1 | ||||
XM_011545316.2 | Intron | XP_011543618.1 | ||||
XM_011545317.2 | Intron | XP_011543619.1 | ||||
XM_011545318.2 | Intron | XP_011543620.1 | ||||
XM_011545319.1 | Intron | XP_011543621.1 | ||||
XM_017018448.1 | Intron | XP_016873937.1 | ||||
XM_017018449.1 | Intron | XP_016873938.1 | ||||
XM_017018450.1 | Intron | XP_016873939.1 |