Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZC3H4 - zinc finger CCCH-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015168.1 | 4292 | UTR 3 | NP_055983.1 | |||
XM_005258676.3 | 4292 | Intron | XP_005258733.1 | |||
XM_005258677.4 | 4292 | Intron | XP_005258734.1 | |||
XM_005258678.2 | 4292 | Intron | XP_005258735.1 | |||
XM_006723113.3 | 4292 | Intron | XP_006723176.1 | |||
XM_011526668.2 | 4292 | Intron | XP_011524970.1 | |||
XM_011526669.2 | 4292 | Intron | XP_011524971.1 | |||
XM_011526670.2 | 4292 | Intron | XP_011524972.1 | |||
XM_017026529.1 | 4292 | Intron | XP_016882018.1 | |||
XM_017026530.1 | 4292 | UTR 3 | XP_016882019.1 | |||
XM_017026531.1 | 4292 | UTR 3 | XP_016882020.1 |
Set Membership: |
HapMap |