Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607103 | ||||||||||||||||||||
Literature Links: |
ATE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATE1 - arginyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001976.2 | 1650 | UTR 3 | NP_001001976.1 | |||
NM_001288734.1 | 1650 | UTR 3 | NP_001275663.1 | |||
NM_001288735.1 | 1650 | UTR 3 | NP_001275664.1 | |||
NM_001288736.1 | 1650 | UTR 3 | NP_001275665.1 | |||
NM_007041.3 | 1650 | UTR 3 | NP_008972.2 | |||
XM_005269458.2 | 1650 | UTR 3 | XP_005269515.1 | |||
XM_005269459.3 | 1650 | UTR 3 | XP_005269516.1 | |||
XM_017015597.1 | 1650 | UTR 3 | XP_016871086.1 | |||
XM_017015598.1 | 1650 | UTR 3 | XP_016871087.1 | |||
XM_017015599.1 | 1650 | UTR 3 | XP_016871088.1 | |||
XM_017015600.1 | 1650 | UTR 3 | XP_016871089.1 | |||
XM_017015601.1 | 1650 | Intron | XP_016871090.1 | |||
XM_017015602.1 | 1650 | UTR 3 | XP_016871091.1 | |||
XM_017015603.1 | 1650 | Intron | XP_016871092.1 | |||
XM_017015604.1 | 1650 | UTR 3 | XP_016871093.1 |