Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607103 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATE1 - arginyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001976.2 | 1675 | UTR 3 | NP_001001976.1 | |||
NM_001288734.1 | 1675 | UTR 3 | NP_001275663.1 | |||
NM_001288735.1 | 1675 | UTR 3 | NP_001275664.1 | |||
NM_001288736.1 | 1675 | UTR 3 | NP_001275665.1 | |||
NM_007041.3 | 1675 | UTR 3 | NP_008972.2 | |||
XM_005269458.2 | 1675 | UTR 3 | XP_005269515.1 | |||
XM_005269459.3 | 1675 | UTR 3 | XP_005269516.1 | |||
XM_017015597.1 | 1675 | UTR 3 | XP_016871086.1 | |||
XM_017015598.1 | 1675 | UTR 3 | XP_016871087.1 | |||
XM_017015599.1 | 1675 | UTR 3 | XP_016871088.1 | |||
XM_017015600.1 | 1675 | UTR 3 | XP_016871089.1 | |||
XM_017015601.1 | 1675 | Intron | XP_016871090.1 | |||
XM_017015602.1 | 1675 | UTR 3 | XP_016871091.1 | |||
XM_017015603.1 | 1675 | Intron | XP_016871092.1 | |||
XM_017015604.1 | 1675 | UTR 3 | XP_016871093.1 |
Set Membership: |
HapMap |