Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607352 MIM: 603612 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHOBTB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RHOBTB2 - Rho related BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF10B - TNF receptor superfamily member 10b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003842.4 | Intron | NP_003833.4 | ||||
NM_147187.2 | Intron | NP_671716.2 | ||||
XM_017013944.1 | Intron | XP_016869433.1 |
Set Membership: |
HapMap |