Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQCE PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IQCE - IQ motif containing E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287499.1 | Intron | NP_001274428.1 | ||||
NM_001287500.1 | Intron | NP_001274429.1 | ||||
NM_001287501.1 | Intron | NP_001274430.1 | ||||
NM_001287502.1 | Intron | NP_001274431.1 | ||||
NM_152558.4 | Intron | NP_689771.3 | ||||
XM_006715676.2 | Intron | XP_006715739.1 | ||||
XM_011515242.1 | Intron | XP_011513544.1 | ||||
XM_011515243.1 | Intron | XP_011513545.1 | ||||
XM_011515244.2 | Intron | XP_011513546.1 | ||||
XM_017011902.1 | Intron | XP_016867391.1 | ||||
XM_017011903.1 | Intron | XP_016867392.1 |