Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGAGGCTTAAACTAACCTAAA[C/T]CAATTGGAGATCAATACAGCCAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609091 MIM: 612325 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO9 - F-box protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012347.4 | Intron | NP_036479.1 | ||||
NM_033480.2 | Intron | NP_258441.1 | ||||
NM_033481.3 | Intron | NP_258442.2 | ||||
XM_005248995.4 | Intron | XP_005249052.1 | ||||
XM_005248996.4 | Intron | XP_005249053.1 |
ICK - intestinal cell (MAK-like) kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014920.3 | Intron | NP_055735.1 | ||||
NM_016513.4 | Intron | NP_057597.2 | ||||
XM_011514419.1 | Intron | XP_011512721.1 | ||||
XM_011514420.2 | Intron | XP_011512722.1 | ||||
XM_011514421.1 | Intron | XP_011512723.1 | ||||
XM_017010485.1 | Intron | XP_016865974.1 | ||||
XM_017010486.1 | Intron | XP_016865975.1 | ||||
XM_017010487.1 | Intron | XP_016865976.1 | ||||
XM_017010488.1 | Intron | XP_016865977.1 | ||||
XM_017010489.1 | Intron | XP_016865978.1 | ||||
XM_017010490.1 | Intron | XP_016865979.1 | ||||
XM_017010491.1 | Intron | XP_016865980.1 | ||||
XM_017010492.1 | Intron | XP_016865981.1 |
LOC105375092 - uncharacterized LOC105375092 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |