Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608774 MIM: 126450 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKK1 - ankyrin repeat and kinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178510.1 | 3949 | Missense Mutation | CCC,CTC | P,L 596 | NP_848605.1 | |
XM_011542736.2 | 3949 | Missense Mutation | CCC,CTC | P,L 607 | XP_011541038.1 | |
XM_011542737.2 | 3949 | Missense Mutation | CCC,CTC | P,L 597 | XP_011541039.1 | |
XM_011542738.2 | 3949 | Missense Mutation | CCC,CTC | P,L 533 | XP_011541040.1 | |
XM_017017475.1 | 3949 | Missense Mutation | CCC,CTC | P,L 606 | XP_016872964.1 |
DRD2 - dopamine receptor D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |