Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613143 MIM: 612066 | |||||||||||||||||||||||
Literature Links: |
DTX3L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DTX3L - deltex E3 ubiquitin ligase 3L | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARP15 - poly(ADP-ribose) polymerase family member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113523.2 | 982 | Intron | NP_001106995.1 | |||
NM_001308320.1 | 982 | Intron | NP_001295249.1 | |||
NM_001308321.1 | 982 | Intron | NP_001295250.1 | |||
NM_152615.2 | 982 | Intron | NP_689828.1 | |||
XM_005247159.3 | 982 | UTR 5 | XP_005247216.1 | |||
XM_005247160.3 | 982 | UTR 5 | XP_005247217.1 | |||
XM_011512475.2 | 982 | UTR 5 | XP_011510777.1 | |||
XM_011512476.2 | 982 | UTR 5 | XP_011510778.1 | |||
XM_011512477.2 | 982 | UTR 5 | XP_011510779.1 | |||
XM_011512478.2 | 982 | Intron | XP_011510780.1 | |||
XM_011512479.2 | 982 | Intron | XP_011510781.1 | |||
XM_011512480.2 | 982 | UTR 5 | XP_011510782.1 | |||
XM_017005791.1 | 982 | UTR 5 | XP_016861280.1 | |||
XM_017005792.1 | 982 | Intron | XP_016861281.1 |