Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF250 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF250 - zinc finger protein 250 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109689.3 | 4659 | UTR 3 | NP_001103159.1 | |||
NM_021061.4 | 4659 | UTR 3 | NP_066405.1 | |||
XM_005272327.1 | 4659 | Intron | XP_005272384.1 | |||
XM_005272328.3 | 4659 | Intron | XP_005272385.1 | |||
XM_006716612.3 | 4659 | UTR 3 | XP_006716675.1 | |||
XM_006716613.3 | 4659 | UTR 3 | XP_006716676.1 | |||
XM_011517209.1 | 4659 | Intron | XP_011515511.1 |