Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615898 MIM: 607077 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFAF7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFAF7 - NADH:ubiquinone oxidoreductase complex assembly factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083946.1 | 4737 | Intron | NP_001077415.1 | |||
NM_144736.4 | 4737 | Intron | NP_653337.1 | |||
XM_005264410.1 | 4737 | Intron | XP_005264467.1 | |||
XM_005264411.1 | 4737 | Intron | XP_005264468.1 | |||
XM_011532956.1 | 4737 | Intron | XP_011531258.1 |
PRKD3 - protein kinase D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005813.4 | 4737 | UTR 3 | NP_005804.1 | |||
XM_005264237.3 | 4737 | UTR 3 | XP_005264294.1 | |||
XM_005264238.3 | 4737 | UTR 3 | XP_005264295.1 | |||
XM_017003741.1 | 4737 | UTR 3 | XP_016859230.1 | |||
XM_017003742.1 | 4737 | UTR 3 | XP_016859231.1 | |||
XM_017003743.1 | 4737 | UTR 3 | XP_016859232.1 | |||
XM_017003744.1 | 4737 | UTR 3 | XP_016859233.1 |
Set Membership: |
HapMap |