Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608172 MIM: 611043 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHDDS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DHDDS - dehydrodolichyl diphosphate synthase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243564.1 | 1 | UTR 5 | NP_001230493.1 | |||
NM_001243565.1 | 1 | UTR 5 | NP_001230494.1 | |||
NM_001319959.1 | 1 | UTR 5 | NP_001306888.1 | |||
NM_024887.3 | 1 | UTR 5 | NP_079163.2 | |||
NM_205861.2 | 1 | UTR 5 | NP_995583.1 | |||
XM_006710912.2 | 1 | Intron | XP_006710975.1 | |||
XM_006710913.2 | 1 | Intron | XP_006710976.1 | |||
XM_006710914.2 | 1 | Intron | XP_006710977.1 | |||
XM_006710915.2 | 1 | Intron | XP_006710978.1 | |||
XM_006710916.2 | 1 | Intron | XP_006710979.1 | |||
XM_006710917.2 | 1 | Intron | XP_006710980.1 | |||
XM_006710918.2 | 1 | Intron | XP_006710981.1 | |||
XM_011542183.2 | 1 | Intron | XP_011540485.1 | |||
XM_011542184.2 | 1 | Intron | XP_011540486.1 | |||
XM_011542185.2 | 1 | Intron | XP_011540487.1 | |||
XM_011542186.2 | 1 | Intron | XP_011540488.1 | |||
XM_017002379.1 | 1 | Intron | XP_016857868.1 | |||
XM_017002380.1 | 1 | Intron | XP_016857869.1 | |||
XM_017002381.1 | 1 | Intron | XP_016857870.1 | |||
XM_017002382.1 | 1 | Intron | XP_016857871.1 | |||
XM_017002383.1 | 1 | Intron | XP_016857872.1 | |||
XM_017002384.1 | 1 | Intron | XP_016857873.1 |
LIN28A - lin-28 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |