Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608944 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FREM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FREM1 - FRAS1 related extracellular matrix 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177704.1 | 8023 | UTR 3 | NP_001171175.1 | |||
NM_144966.5 | 8023 | UTR 3 | NP_659403.4 | |||
XM_005251382.3 | 8023 | UTR 3 | XP_005251439.1 | |||
XM_005251384.4 | 8023 | UTR 3 | XP_005251441.1 | |||
XM_006716729.3 | 8023 | UTR 3 | XP_006716792.1 | |||
XM_011517758.2 | 8023 | UTR 3 | XP_011516060.1 | |||
XM_017014316.1 | 8023 | UTR 3 | XP_016869805.1 | |||
XM_017014317.1 | 8023 | UTR 3 | XP_016869806.1 | |||
XM_017014318.1 | 8023 | UTR 3 | XP_016869807.1 | |||
XM_017014319.1 | 8023 | UTR 3 | XP_016869808.1 | |||
XM_017014320.1 | 8023 | UTR 3 | XP_016869809.1 | |||
XM_017014321.1 | 8023 | UTR 3 | XP_016869810.1 | |||
XM_017014322.1 | 8023 | UTR 3 | XP_016869811.1 | |||
XM_017014323.1 | 8023 | UTR 3 | XP_016869812.1 | |||
XM_017014324.1 | 8023 | UTR 3 | XP_016869813.1 | |||
XM_017014325.1 | 8023 | UTR 3 | XP_016869814.1 | |||
XM_017014326.1 | 8023 | UTR 3 | XP_016869815.1 | |||
XM_017014327.1 | 8023 | UTR 3 | XP_016869816.1 | |||
XM_017014328.1 | 8023 | Intron | XP_016869817.1 | |||
XM_017014329.1 | 8023 | Intron | XP_016869818.1 | |||
XM_017014330.1 | 8023 | Intron | XP_016869819.1 |