Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603267 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAPN15 - calpain 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005632.2 | Intron | NP_005623.1 | ||||
XM_011522620.2 | Intron | XP_011520922.1 | ||||
XM_011522621.2 | Intron | XP_011520923.1 | ||||
XM_011522622.1 | Intron | XP_011520924.1 | ||||
XM_011522623.2 | Intron | XP_011520925.1 | ||||
XM_011522624.2 | Intron | XP_011520926.1 | ||||
XM_011522625.2 | Intron | XP_011520927.1 | ||||
XM_011522626.2 | Intron | XP_011520928.1 | ||||
XM_011522627.2 | Intron | XP_011520929.1 | ||||
XM_011522628.2 | Intron | XP_011520930.1 | ||||
XM_011522629.2 | Intron | XP_011520931.1 | ||||
XM_011522630.2 | Intron | XP_011520932.1 | ||||
XM_011522631.2 | Intron | XP_011520933.1 | ||||
XM_011522632.2 | Intron | XP_011520934.1 | ||||
XM_017023596.1 | Intron | XP_016879085.1 |
MIR3176 - microRNA 3176 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5587 - microRNA 5587 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |