Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603651 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRPC4 - transient receptor potential cation channel subfamily C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135955.1 | 2744 | Intron | NP_001129427.1 | |||
NM_001135956.1 | 2744 | Intron | NP_001129428.1 | |||
NM_001135957.1 | 2744 | Intron | NP_001129429.1 | |||
NM_001135958.1 | 2744 | Intron | NP_001129430.1 | |||
NM_003306.1 | 2744 | Intron | NP_003297.1 | |||
NM_016179.2 | 2744 | Intron | NP_057263.1 | |||
XM_011535206.1 | 2744 | UTR 3 | XP_011533508.1 | |||
XM_017020723.1 | 2744 | UTR 3 | XP_016876212.1 | |||
XM_017020724.1 | 2744 | Intron | XP_016876213.1 |
Set Membership: |
HapMap |